1-(2-fluoro-4-nitrophenyl)-4-methylpiperazine
Supplier: Apollo Scientific
Synonyms: 1-(2-Fluoro-4-nitrophenyl)-4-methylpiperazine
PC10263-1G PC10263-25G PC10263-5G
APOSPC10263-1GEA 30.6 EUR
APOSPC10263-1G APOSPC10263-25G APOSPC10263-5G
1-(2-fluoro-4-nitrophenyl)-4-methylpiperazine
1-(2-fluoro-4-nitrophenyl)-4-methylpiperazine
Formula: C₁₁H₁₄FN₃O₂
Melting Pt: 71…72 °C
Storage Temperature: Ambient
MDL Number: MFCD02633484
CAS Number: 221198-29-4