5-Fluoro-2H-3,1-benzoxazine-2,4(1H)-dione 96%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Fluoro-1H-benzo[d][1,3]oxazine-2,4-dione, 6-Fluoroisatoic anhydride
PC10266-1G PC10266-25G PC10266-5G
APOSPC10266-1GEA 59.6 EUR
APOSPC10266-1G APOSPC10266-25G APOSPC10266-5G
5-Fluoro-2H-3,1-benzoxazine-2,4(1H)-dione 96%
5-Fluoro-2H-3,1-benzoxazine-2,4(1H)-dione
Formula: C₈H₄FNO₃
Melting Pt: 230 °C
Density: 1.502 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00112979
CAS Number: 78755-94-9