1-[1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-2,2,2-trifluoro-1-ethanone
Supplier: Apollo Scientific
PC10267-1G PC10267-500MG PC10267-5G
APOSPC10267-1GEA 172 EUR
APOSPC10267-1G APOSPC10267-500MG APOSPC10267-5G
1-[1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-2,2,2-trifluoro-1-ethanone
1-[1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-2,2,2-trifluoro-1-ethanone
Formula: C₁₄H₁₁BrF₃NO
MW: 346.15 g/mol
Melting Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00268899
CAS Number: 95337-69-2