3-Fluoro-1-iodopropane 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Fluoro-3-iodopropane
PC1027-100G PC1027-25G PC1027-5G
APOSPC1027-100GEA 433 EUR
APOSPC1027-100G APOSPC1027-25G APOSPC1027-5G
3-Fluoro-1-iodopropane 98%
3-Fluoro-1-iodopropane
Formula: C₃H₆FI
MW: 187.982 g/mol
Boiling Pt: 68…69 °C/95mmHg
Flash Pt: 34 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07784197
CAS Number: 462-40-8
UN: 1993
ADR: 3,III