2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic acid
Supplier: Apollo Scientific
Warning
PC10287-1G PC10287-250MG PC10287-5G
APOSPC10287-1GEA 55.3 EUR
APOSPC10287-1G APOSPC10287-250MG APOSPC10287-5G
2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic acid
2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic acid
Formula: C₆H₄F₃NO₂S
MW: 211.1600 g/mol
Melting Pt: 186…187 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00173295
CAS Number: 117724-63-7