Pentafluorophenyl-4-chlorobenzenesulphonate
Supplier: Apollo Scientific
PC10308-1G PC10308-5G
APOSPC10308-1GEA 201 EUR
APOSPC10308-1G APOSPC10308-5G
Pentafluorophenyl-4-chlorobenzenesulphonate
Pentafluorophenyl 4-chlorobenzenesulphonate
Formula: C₁₂H₄ClF₅O₃S
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD05975123
CAS Number: 161759-99-5