Pentafluorophenyl-3-(trifluoromethyl)benzenesulphonate
Supplier: Apollo Scientific
PC10310-1G PC10310-500MG PC10310-5G
APOSPC10310-1GEA 275 EUR
APOSPC10310-1G APOSPC10310-500MG APOSPC10310-5G
Pentafluorophenyl-3-(trifluoromethyl)benzenesulphonate
Pentafluorophenyl 3-(trifluoromethyl)benzenesulphonate
Formula: C₁₃H₄F₈O₃S
Storage Temperature: Ambient
MDL Number: MFCD05975128
CAS Number: 885950-36-7