Pentafluorophenyl-4-fluorobenzenesulphonate
Supplier: Apollo Scientific
PC10324-1G PC10324-500MG PC10324-5G
APOSPC10324-1GEA 306 EUR
APOSPC10324-1G APOSPC10324-500MG APOSPC10324-5G
Pentafluorophenyl-4-fluorobenzenesulphonate
Pentafluorophenyl 4-fluorobenzenesulphonate
Formula: C₁₂H₄F₆O₃S
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD05975136
CAS Number: 885950-58-3