1-Bromo-3-fluoro-2-iodobenzene 99%
Supplier: Apollo Scientific
Warning
PC10331-5G PC10331-100G PC10331-25G
APOSPC10331-5GEA 21.9 EUR
APOSPC10331-5G APOSPC10331-100G APOSPC10331-25G
1-Bromo-3-fluoro-2-iodobenzene 99%
1-Bromo-3-fluoro-2-iodobenzene
Formula: C₆H₃BrFI
MW: 300.8949 g/mol
Boiling Pt: 108…109 °C/10mmHg
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD07368675
CAS Number: 450412-29-0