4-Hydroxy-3-(trifluoromethyl)benzonitrile
Supplier: Apollo Scientific
Warning
PC10355-500MG PC10355-5G PC10355-25G
APOSPC10355-500MGEA 16.1 EUR
APOSPC10355-500MG APOSPC10355-5G APOSPC10355-25G
4-Hydroxy-3-(trifluoromethyl)benzonitrile
4-Hydroxy-3-(trifluoromethyl)benzonitrile
Formula: C₈H₄F₃NO
MW: 187.12 g/mol
Melting Pt: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD06797882
CAS Number: 124811-71-8