Ethyl-7-amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylate
Supplier: Apollo Scientific
Warning
PC10359-1G PC10359-250MG
APOSPC10359-1GEA 279 EUR
APOSPC10359-1G APOSPC10359-250MG
Ethyl-7-amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylate
Ethyl-7-amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylate
Formula: C₁₇H₁₅F₃N₄O₂
MW: 364.33 g/mol
Melting Pt: 179…181 °C
Storage Temperature: Ambient
MDL Number: MFCD06200917
CAS Number: 886762-57-8