5-Bromo-2-fluoropyridine 98%
Supplier: Apollo Scientific
Warning
PC10362-100G PC10362-25G PC10362-5G PC10362-500G
APOSPC10362-100GEA 65.4 EUR
APOSPC10362-100G APOSPC10362-25G APOSPC10362-5G APOSPC10362-500G
5-Bromo-2-fluoropyridine 98%
5-Bromo-2-fluoropyridine
Formula: C₅H₃BrFN
MW: 175.99 g/mol
Boiling Pt: 170…173 °C
Density: 1.71 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD01863742
CAS Number: 766-11-0