1-Methyl-5-(phenylsulphanyl)-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Supplier: Apollo Scientific
PC10371-1G PC10371-500MG PC10371-5G
APOSPC10371-1GEA 405 EUR
APOSPC10371-1G APOSPC10371-500MG APOSPC10371-5G
1-Methyl-5-(phenylsulphanyl)-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
1-Methyl-5-(phenylsulphanyl)-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Formula: C₁₂H₉F₃N₂OS
MW: 286.28 g/mol
Melting Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00140823
CAS Number: 321533-77-1