β-Chloro-4'-fluoropropiophenone
Supplier: Apollo Scientific
Warning
Synonyms: 3-Chloro-1-(4-fluorophenyl)propan-1-one, 3-Chloro-4′-fluoropropiophenone, 3-Chloro-4'-fluoropropiophenone
PC10384-100G PC10384-25G PC10384-5G
APOSPC10384-100GEA 234 EUR
APOSPC10384-100G APOSPC10384-25G APOSPC10384-5G
β-Chloro-4'-fluoropropiophenone
β-Chloro-4'-fluoropropiophenone
Formula: C₉H₈ClFO
MW: 186.61 g/mol
Boiling Pt: 100…103 °C/3mmHg
Melting Pt: 47…49 °C
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00000991
CAS Number: 347-93-3
EINECS: 206-475-2