2-(2,2,2-Trifluoroacetyl)thiophene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Trifluoroacetyl)thiophene
PC10396-1G PC10396-5G PC10396-250MG
APOSPC10396-1GEA 98.9 EUR
APOSPC10396-1G APOSPC10396-5G APOSPC10396-250MG
2-(2,2,2-Trifluoroacetyl)thiophene 98%
2-(2,2,2-Trifluoroacetyl)thiophene
Formula: C₆H₅ClOS
MW: 180.15 g/mol
Boiling Pt: 163…165 °C
Density: 1.403 g/cm³ (25 °C)
Flash Pt: 59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041427
CAS Number: 651-70-7
UN: 1993
ADR: 3,III