5-Fluoro-2-methoxypyridine 97%
Supplier: Apollo Scientific
Warning
PC10417-1G PC10417-5G PC10417-10G
APOSPC10417-1GEA 21.9 EUR
APOSPC10417-1G APOSPC10417-5G APOSPC10417-10G
5-Fluoro-2-methoxypyridine 97%
5-Fluoro-2-methoxypyridine
Formula: C₆H₆FNO
MW: 127.12 g/mol
Boiling Pt: 137…40 °C
Flash Pt: <60 °C
Storage Temperature: Freezer
MDL Number: MFCD03095252
CAS Number: 51173-04-7
UN: 1993
ADR: 3 III