4-Bromo-2-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Warning
PC1042-100G PC1042-500G PC1042-50G
APOSPC1042-100GEA 50.9 EUR
APOSPC1042-100G APOSPC1042-500G APOSPC1042-50G
4-Bromo-2-(trifluoromethyl)aniline 98%
4-Bromo-2-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
Boiling Pt: 84…86 °C/5mmHg
Density: 1.71 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064393
CAS Number: 445-02-3
EINECS: 207-150-8