2-Fluoro-5-iodopyridine 98%
Supplier: Apollo Scientific
Warning
PC10421-100G PC10421-25G PC10421-5G
APOSPC10421-100GEA 275 EUR
APOSPC10421-100G APOSPC10421-25G APOSPC10421-5G
2-Fluoro-5-iodopyridine 98%
2-Fluoro-5-iodopyridine
Formula: C₅H₃FIN
MW: 222.99 g/mol
Melting Pt: 35…36 °C
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD03095301
CAS Number: 171197-80-1