2-Bromo-5-fluoro-6-methylpyridine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-5-fluoro-6-picoline
PC10422-25G PC10422-5G PC10422-10G
APOSPC10422-25GEA 78.3 EUR
APOSPC10422-25G APOSPC10422-5G APOSPC10422-10G
2-Bromo-5-fluoro-6-methylpyridine 98%
2-Bromo-5-fluoro-6-methylpyridine
Formula: C₆H₅BrFN
MW: 190.01 g/mol
Melting Pt: 63…65 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD03095064
CAS Number: 374633-38-2