2-Bromo-2',4',5'-trifluoroacetophenone 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,4,5-Trifluorophenacyl bromide
PC10423-1G PC10423-5G
APOSPC10423-1GEA 244 EUR
APOSPC10423-1G APOSPC10423-5G
2-Bromo-2',4',5'-trifluoroacetophenone 98%
2-Bromo-2',4',5'-trifluoroacetophenone
Formula: C₈H₄BrF₃O
Melting Pt: 58…60 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368689
CAS Number: 193977-34-3
UN: 1759
ADR: 8,III