3-Fluoro-4-(trifluoromethoxy)phenol
Supplier: Apollo Scientific
Warning
PC10448-1G PC10448-5G PC10448-25G
APOSPC10448-1GEA 21.9 EUR
APOSPC10448-1G APOSPC10448-5G APOSPC10448-25G
3-Fluoro-4-(trifluoromethoxy)phenol
3-Fluoro-4-(trifluoromethoxy)phenol
Formula: C₇H₄F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD06660342
CAS Number: 177596-38-2