5-Bromo-2-fluoro-4-methylaniline
Supplier: Apollo Scientific
Warning
PC10501-1G PC10501-5G PC10501-25G
APOSPC10501-1GEA 34.8 EUR
APOSPC10501-1G APOSPC10501-5G APOSPC10501-25G
5-Bromo-2-fluoro-4-methylaniline
5-Bromo-2-fluoro-4-methylaniline
Formula: C₇H₇BrFN
MW: 204.0400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11504886
CAS Number: 945244-29-1