2-Amino-3,5-difluoropyridine
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Difluoro-2-pyridylamine, 3,5-Difluoropyridin-2-amine
PC10505-25G
APOSPC10505-25GEA 306 EUR
APOSPC10505-25G
2-Amino-3,5-difluoropyridine
2-Amino-3,5-difluoropyridine
Formula: C₅H₄F₂N₂
Melting Pt: 52…59 °C
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD03788760
CAS Number: 732306-31-9