3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane
Supplier: Apollo Scientific
Synonyms: [3,5-Bis(trifluoromethyl)phenyl]chloro(dimethyl)silane
PC10507-1G PC10507-5G
APOSPC10507-1GEA 118 EUR
APOSPC10507-1G APOSPC10507-5G
3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane
3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane
Formula: C₁₀H₉ClF₆Si
Boiling Pt: 95…96 °C/30mmHg
Storage Temperature: Ambient
MDL Number: MFCD01862186
CAS Number: 732306-23-9