tert-Butyl-6-fluoro-1H-pyrazolo[3,4-b]pyridine-3-carboxylate 97%
Supplier: Apollo Scientific
Warning
PC1051-250MG PC1051-50MG
APOSPC1051-250MGEA 174 EUR
APOSPC1051-250MG APOSPC1051-50MG
tert-Butyl-6-fluoro-1H-pyrazolo[3,4-b]pyridine-3-carboxylate 97%
tert-Butyl-6-fluoro-1H-pyrazolo[3,4-b]pyridine-3-carboxylate
Formula: C₁₁H₁₂FN₃O₂
MW: 237.24 g/mol
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD06797502
CAS Number: 155601-71-1