2-Bromo-5-(trifluoromethyl)phenylacetonitrile
Supplier: Apollo Scientific
Warning
PC10515-1G PC10515-5G
APOSPC10515-1GEA 139 EUR
APOSPC10515-1G APOSPC10515-5G
2-Bromo-5-(trifluoromethyl)phenylacetonitrile
2-Bromo-5-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅BrF₃N
MW: 264.04 g/mol
Boiling Pt: 144…146 °C/15mmHg
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD04973763
CAS Number: 732306-26-2
UN: 3439
ADR: 6.1 III