2,3,5-Trifluoropyridine 98%
Supplier: Apollo Scientific
Warning
PC10551-25G PC10551-5G PC10551-100G
APOSPC10551-25GEA 137 EUR
APOSPC10551-25G APOSPC10551-5G APOSPC10551-100G
2,3,5-Trifluoropyridine 98%
2,3,5-Trifluoropyridine
Formula: C₅H₂F₃N
Boiling Pt: 102 °C
Density: 1.4 g/cm³ (25 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD03001162
CAS Number: 76469-41-5
UN: 1993
ADR: 3,III