2,4,6-Trifluoropyridine 97%
Supplier: Apollo Scientific
|
Danger
|
PC10553-1G
PC10553-5G
APOSPC10553-1GEA
96.3
EUR
APOSPC10553-1G
APOSPC10553-5G
2,4,6-Trifluoropyridine 97%
2,4,6-Trifluoropyridine
Formula:
C₅H₂F₃N Boiling Pt: 97…99 °C Density: 1.499 g/cm³ (25 °C) Flash Pt: 17 °C Storage Temperature: Ambient |
MDL Number:
MFCD03001161 CAS Number: 3512-17-2 UN: 2924 ADR: 3+8,II |