2,4-Difluoropyrimidine 98%
Supplier: Apollo Scientific
Danger
PC1056-5G PC1056-10G PC1056-1G
APOSPC1056-5GEA 299 EUR
APOSPC1056-5G APOSPC1056-10G APOSPC1056-1G
2,4-Difluoropyrimidine 98%
2,4-Difluoropyrimidine
Formula: C₄H₂F₂N₂
MW: 116.07 g/mol
Boiling Pt: 118…120 °C
Density: 1.359 g/cm³ (25 °C)
Flash Pt: 41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972830
CAS Number: 2802-61-1
UN: 1993
ADR: 3,III