5-Methyl-3-[3-(trifluoromethyl)phenyl]-1H-pyrazole
Supplier: Apollo Scientific
Warning
PC10564-1G PC10564-500MG PC10564-5G
APOSPC10564-1GEA 350 EUR
APOSPC10564-1G APOSPC10564-500MG APOSPC10564-5G
5-Methyl-3-[3-(trifluoromethyl)phenyl]-1H-pyrazole
5-Methyl-3-[3-(trifluoromethyl)phenyl]-1H-pyrazole
Formula: C₁₁H₉F₃N₂
MW: 226.2 g/mol
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD06659570
CAS Number: 521267-21-0