Pentafluorophenyl-2,3-dichlorobenzenesulphonate
Supplier: Apollo Scientific
PC10568-1G PC10568-5G
APOSPC10568-1GEA 201 EUR
APOSPC10568-1G APOSPC10568-5G
Pentafluorophenyl-2,3-dichlorobenzenesulphonate
Pentafluorophenyl 2,3-dichlorobenzenesulphonate
Formula: C₁₂H₃Cl₂F₅O₃S
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD06659539
CAS Number: 885949-54-2