Pentafluorophenyl-2,4-dichlorobenzenesulphonate
Supplier: Apollo Scientific
PC10569-1G PC10569-5G
APOSPC10569-1GEA 183 EUR
APOSPC10569-1G APOSPC10569-5G
Pentafluorophenyl-2,4-dichlorobenzenesulphonate
Pentafluorophenyl 2,4-dichlorobenzenesulphonate
Formula: C₁₂H₃Cl₂F₅O₃S
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD06659543
CAS Number: 885949-57-5