Pentafluorophenyl-5-(dimethylamino)-1-naphthalenesulphonate
Supplier: Apollo Scientific
PC10571-1G PC10571-500MG PC10571-5G
APOSPC10571-1GEA 275 EUR
APOSPC10571-1G APOSPC10571-500MG APOSPC10571-5G
Pentafluorophenyl-5-(dimethylamino)-1-naphthalenesulphonate
Pentafluorophenyl 5-(dimethylamino)-1-naphthalenesulphonate
Formula: C₁₈H₁₂F₅NO₃S
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD06659540
CAS Number: 663175-92-6