1-(2,3-Difluorophenyl)-2-thiourea 97%
Supplier: Apollo Scientific
Warning
Synonyms: N-(2,3-Difluorophenyl)thiourea, 1-(2,3-Difluorophenyl)thiourea, 2,3-Difluorophenylthiourea
PC10573-1G PC10573-5G PC10573-10G
APOSPC10573-1GEA 71.2 EUR
APOSPC10573-1G APOSPC10573-5G APOSPC10573-10G
1-(2,3-Difluorophenyl)-2-thiourea 97%
1-(2,3-Difluorophenyl)-2-thiourea
Formula: (C₆H₃F₂)NC(S)NH₂
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD07368705
CAS Number: 572889-25-9