Methyl-1-trifluoromethyl-1-cyclopropanecarboxylate 95%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 1-(trifluoromethyl)cyclopropanecarboxylate
PC1058-1G PC1058-5G
APOSPC1058-1GEA 143 EUR
APOSPC1058-1G APOSPC1058-5G
Methyl-1-trifluoromethyl-1-cyclopropanecarboxylate 95%
Methyl 1-trifluoromethyl-1-cyclopropanecarboxylate
Formula: C₆H₇F₃O₂
Boiling Pt: 64…66 °C
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD06801321
CAS Number: 208242-25-5
UN: 3272
ADR: 3,III