5-Fluoro-2-methoxybenzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 5-Fluoro-2-methoxybenzene-1-sulphonyl chloride
PC10595-1G PC10595-5G
APOSPC10595-1GEA 21.9 EUR
APOSPC10595-1G APOSPC10595-5G
5-Fluoro-2-methoxybenzenesulphonyl chloride
5-Fluoro-2-methoxybenzenesulphonyl chloride
Formula: C₇H₆ClFO₃S
MW: 224.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03618459
CAS Number: 67475-56-3
UN: 3261
ADR: 8,III