4-Bromotetrafluorophenol 96%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-2,3,5,6-tetrafluorophenol
PC10598-10G PC10598-1G PC10598-5G
APOSPC10598-10GEA 284 EUR
APOSPC10598-10G APOSPC10598-1G APOSPC10598-5G
4-Bromotetrafluorophenol 96%
4-Bromotetrafluorophenol
Formula: C₆HBrF₄O
MW: 244.98 g/mol
Boiling Pt: 76…77 °C/4mmHg
Melting Pt: 38…39 °C
Storage Temperature: Ambient
MDL Number: MFCD06246024
CAS Number: 1998-61-4