2-Chloro-5-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Amino-4-chloro-α,α,α-trifluortoluene, 3-Amino-4-chloro-α,α,α-trifluorotoluene, 6-Chloro-α,α,α-trifluoro-m-toluidine, 3-Amino-4-chlorobenzotrifluoride
PC1060-100G PC1060-500G
APOSPC1060-100GEA 21.9 EUR
APOSPC1060-100G APOSPC1060-500G
2-Chloro-5-(trifluoromethyl)aniline 98%
2-Chloro-5-(trifluoromethyl)aniline
Formula: C₇H₅ClF₃N
MW: 195.57 g/mol
Boiling Pt: 82…83 °C/9mmHg
Melting Pt: 44477 °C
Density: 1.428 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00007673
CAS Number: 121-50-6
EINECS: 204-475-7
UN: 2810
ADR: 6.1,III