Methyl bromodifluoroacetate
Supplier: Apollo Scientific
Danger
Synonyms: Bromodifluoroacetic Acid Methyl Ester
PC10611-5G PC10611-100G
APOSPC10611-5GEA 31.9 EUR
APOSPC10611-5G APOSPC10611-100G
Methyl bromodifluoroacetate
Methyl bromodifluoroacetate
Formula: C₃H₃BrF₂O₂
MW: 188.96 g/mol
Boiling Pt: 96 °C
Density: 1.72 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD06248118
CAS Number: 683-98-7
UN: 2924
ADR: 3+8,II