2-Trifluoromethyl acrylonitrile
Supplier: Apollo Scientific
Danger
PC10628-5G PC10628-10G
APOSPC10628-5GEA 364 EUR
APOSPC10628-5G APOSPC10628-10G
2-Trifluoromethyl acrylonitrile
2-Trifluoromethyl acrylonitrile
Formula: C₄H₂F₃N
MW: 121.06 g/mol
Boiling Pt: 76…77 °C
Density: 1.1753 g/cm³ (25 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD01673877
CAS Number: 381-84-0
UN: 1992
ADR: 3+6.1,II