4-Chloro-3-(trifluoromethyl)aniline 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Amino-2-chloro-α,α,α-trifluorotoluene, 1-Amino-4-chloro-3-(trifluoromethyl)benzene, 4-Chloro-α,α,α-trifluoro-m-toluidine, 5-Amino-2-chlorobenzotrifluoride
PC1070-25G PC1070-100G
APOSPC1070-25GEA 21.9 EUR
APOSPC1070-25G APOSPC1070-100G
4-Chloro-3-(trifluoromethyl)aniline 97%
4-Chloro-3-(trifluoromethyl)aniline
Formula: C₇H₅ClF₃N
Boiling Pt: 85 °C/3mmHg
Melting Pt: 35…38 °C
Flash Pt: 134 °C
Storage Temperature: Ambient
MDL Number: MFCD00007846
CAS Number: 320-51-4
EINECS: 206-277-6
UN: 2811
ADR: 6.1,III