2-Amino-6-fluorobenzonitrile 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Cyano-3-fluoroaniline
PC1071Q-100G PC1071Q-1KG PC1071Q-500G
APOSPC1071Q-100GEA 34.8 EUR
APOSPC1071Q-100G APOSPC1071Q-1KG APOSPC1071Q-500G
2-Amino-6-fluorobenzonitrile 98%
2-Amino-6-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.13 g/mol
Boiling Pt: 272
Melting Pt: 125…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00015477
CAS Number: 77326-36-4
UN: 3439
ADR: 6.1 III