2-Fluoro-6-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-α,α,α,3-tetrafluorotoluene, 2-Amino-alpha,alpha,alpha,3-tetrafluorotoluene, 2-Amino-3-fluorobenzotrifluoride, α,α,α,6-Tetrafluoro-o-toluidine
PC1071R-1G PC1071R-5G PC1071R-25G
APOSPC1071R-1GEA 21.9 EUR
APOSPC1071R-1G APOSPC1071R-5G APOSPC1071R-25G
2-Fluoro-6-(trifluoromethyl)aniline 98%
2-Fluoro-6-(trifluoromethyl)aniline
Formula: C₇H₅F₄N
MW: 179.12 g/mol
Boiling Pt: 155 °C
Density: 1.388 g/cm³ (25 °C)
Flash Pt: 53 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00061242
CAS Number: 144851-61-6
UN: 1993
ADR: 3 III