2-Fluoro-3-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Amino-alpha,alpha,alpha,2-tetrafluorotoluene, 3-Amino-2-fluorobenzotrifluoride, α,α,α,2-Tetrafluoro-m-toluidine, 3-Amino-α,α,α,2-tetrafluorotoluene
PC1073-10G PC1073-25G PC1073-100G
APOSPC1073-10GEA 33.5 EUR
APOSPC1073-10G APOSPC1073-25G APOSPC1073-100G
2-Fluoro-3-(trifluoromethyl)aniline 98%
2-Fluoro-3-(trifluoromethyl)aniline
Formula: C₇H₅F₄N
MW: 179.12 g/mol
Boiling Pt: 189 °C
Density: 1.39 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00061241
CAS Number: 123973-25-1
UN: 2810
ADR: 6.1,III