3-Fluoro-5-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Danger
PC1075-5G PC1075-25G
APOSPC1075-5GEA 33.5 EUR
APOSPC1075-5G APOSPC1075-25G
3-Fluoro-5-(trifluoromethyl)aniline 98%
3-Fluoro-5-(trifluoromethyl)aniline
Formula: C₇H₅F₄N
Storage Temperature: Ambient
MDL Number: MFCD00061315
CAS Number: 454-67-1
UN: 2810
ADR: 6.1,III