2-Amino-4-(trifluoromethyl)thiophenol hydrochloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Amino-4-(trifluoromethyl)benzenethiol hydrochloride
PC1077-25G PC1077-5G
APOSPC1077-25GEA 256 EUR
APOSPC1077-25G APOSPC1077-5G
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride 97%
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride
Formula: C₇H₇ClF₃NS
MW: 229.65
Melting Pt: 186…190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042600
CAS Number: 4274-38-8
EINECS: 429-560-8
UN: 1759
ADR: 8 II