3-Bromo-5-(trifluoromethyl)pyridine 98%
Supplier: Apollo Scientific
Danger
PC1078-1G PC1078-25G PC1078-5G
APOSPC1078-1GEA 21.9 EUR
APOSPC1078-1G APOSPC1078-25G APOSPC1078-5G
3-Bromo-5-(trifluoromethyl)pyridine 98%
3-Bromo-5-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃N
MW: 226.00 g/mol
Boiling Pt: 65…66 °C/19mmHg
Melting Pt: 44…46 °C
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD04972700
CAS Number: 436799-33-6
UN: 2811
ADR: 6.1 III