4-[(Trifluoromethyl)thio]aniline 96%
Supplier: Apollo Scientific
Warning
Synonyms: 4-((Trifluoromethyl)thio)aniline
PC1095K-10G PC1095K-100G PC1095K-25G
APOSPC1095K-10GEA 21.9 EUR
APOSPC1095K-10G APOSPC1095K-100G APOSPC1095K-25G
4-[(Trifluoromethyl)thio]aniline 96%
4-[(Trifluoromethyl)thio]aniline
Formula: C₇H₆F₃NS
Boiling Pt: 102…103 °C/8mmHg
Density: 1.351 g/cm³ (25 °C)
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00040926
CAS Number: 372-16-7
EINECS: 206-744-4