3-Amino-2,4,5-trifluorobenzoic acid 97%
Supplier: Apollo Scientific
|
Warning
|
PC1099G-25G
PC1099G-5G
APOSPC1099G-25GEA
475
EUR
APOSPC1099G-25G
APOSPC1099G-5G
3-Amino-2,4,5-trifluorobenzoic acid 97%
3-Amino-2,4,5-trifluorobenzoic acid
Formula:
C₇H₄F₃NO₂ Melting Pt: 133…136 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00077453 CAS Number: 119385-80-7 |