2-Fluoro-5-(trifluoromethoxy)phenylacetic acid 97%
Supplier: Apollo Scientific
|
Warning
|
PC1100-1G
PC1100-5G
APOSPC1100-1GEA
232
EUR
APOSPC1100-1G
APOSPC1100-5G
2-Fluoro-5-(trifluoromethoxy)phenylacetic acid 97%
2-Fluoro-5-(trifluoromethoxy)phenylacetic acid
Formula:
C₉H₆F₄O₃ MW: 238.14 g/mol Melting Pt: 54…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115892 CAS Number: 886497-98-9 |